Assay Design



Gene Transcripts

Gene Symbol
Hsc70-1
Entrez Gene ID
39542
Gene Name
Heat shock protein cognate 1
Gene Aliases
143191_at, BcDNA:RH49358, CG8937, DmHsp70, Dmel\CG8937, Dmel_CG8937, HSC-70, HSC1, HSC70, HSC70-1, HSP7A_DROME, Hsc1, Hsc70, Hsp-c1, Hsp70, dhsc70, dhsc70-1, hsc70
Chromosome Location
Chr.3L: 13974061 - 13977761 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001216
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079339.3 NP_524063.1 2 - 2 1041 57
NM_168568.3 NP_729940.1 3 - 3 1484 57
NM_168569.2 NP_729941.1 4 - 4 1416 57
GenBank mRNA BT001277.1 - 1 - 1 820 57
L01501.1 - 2 - 2 1179 57

View Full Product Details