Assay Design
Gene Transcripts
- Gene Symbol
- tgo
- Entrez Gene ID
-
41084
- Gene Name
- tango
- Gene Aliases
-
ARNT, Arnt, CG11987, DARNT, Dmel\CG11987, Dmel_CG11987, HIF-1, HIF-1beta, HIF1B, HIF1B/ARNT, HIF1b, Hif1, TGO, TGO_CYC, Tango, Tgo, arnt, bHLHe4, dARNT, dHIF-1beta, dTgo, darnt, jay, prd7, tgo/Arnt
- Chromosome Location
-
Chr.3R: 9016773 - 9020022 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0264075
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_169254.2 |
NP_731308.1 |
2 - 2 |
2969 |
146 |
View Full Product Details