Assay Design
Gene Transcripts
- Gene Symbol
- Tgs1
- Entrez Gene ID
-
59260
- Gene Name
- Trimethylguanosine synthase 1
- Gene Aliases
-
0967/13, CG31241, CG44890, CG8045, CT24102, DTL, DTLd, Dmel\CG44890, Dmel_CG44890, Dromel_CG31241_FBtr0083570_dtl_mORF, Dtl, TGS1, dORF, dtl, l(3)S096713, tatl, tgs1
- Chromosome Location
-
Chr.3R: 18255530 - 18257677 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0266195
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_169801.2 |
NP_732316.2 |
2 - 2 |
1960 |
84 |
- Gene Symbol
- moi
- Entrez Gene ID
-
7354473
- Gene Name
- modigliani
- Gene Aliases
-
0967/13, CG31241, CG42350, DTL, DTL/Moi, DTLu, Dmel\CG42350, Dmel_CG42350, Dromel_CG31241_FBtr0083570_dtl_uORF, Dtl uORF, Moi, l(3)CB02140, l(3)S096713, tlu, uORF
- Chromosome Location
-
Chr.3R: 18255530 - 18257677 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261019
View Full Product Details