Assay Design



Gene Transcripts

Gene Symbol
fray
Entrez Gene ID
44233
Gene Name
frayed
Gene Aliases
4624, CG7693, Dmel\CG7693, Dmel_CG7693, Dosr1, Fray, STE20-like/SPAK, dOSR1, dSTRAD, l(3)07551, lag
Chromosome Location
Chr.3R: 18573091 - 18589459 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0023083
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144605.2 NP_001138077.1 2 - 2 2067 78
NM_001144606.1 NP_001138078.1 2 - 2 1727 78
NM_001144607.2 NP_001138079.1 2 - 2 1820 78
NM_001300466.1 NP_001287395.1 4 - 4 2020 78
NM_001316520.1 NP_001303449.2 2 - 2 1921 78
NM_001316521.1 NP_001303450.1 3 - 3 1830 78
NM_057960.4 NP_477308.1 3 - 3 2085 78
NM_169823.2 NP_732352.1 2 - 2 1746 78
GenBank mRNA BT003198.1 - 3 - 3 2081 78
BT021945.1 - 2 - 2 2047 78

View Full Product Details