Assay Design



Gene Transcripts

Gene Symbol
bocks
Entrez Gene ID
41162
Gene Name
bocksbeutel
Gene Aliases
Bocks, Bocksbeutel, CG9424, Dmel\CG9424, Dmel_CG9424, emerin
Chromosome Location
Chr.3R: 9589468 - 9591002 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037719
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014611.3 NP_001014611.1 1 - 1 1168 105
GenBank mRNA AJ606681.1 - 1 - 1 1153 105

View Full Product Details