Assay Design
Gene Transcripts
- Gene Symbol
- Peritrophin-15a
- Entrez Gene ID
-
50433
- Gene Name
- CG17814-PA
- Gene Aliases
-
CG17814, CT39472, Dm15a, Dmel\CG17814, Dmel_CG17814, Peritrophin-15A, peritrophin-15a
- Chromosome Location
-
Chr.2L: 8395665 - 8396096 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0040959
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080244.2 |
NP_524983.1 |
2 - 2 |
283 |
64 |
View Full Product Details