Assay Design



Gene Transcripts

Gene Symbol
ome
Entrez Gene ID
44297
Gene Name
omega
Gene Aliases
BEST:LD21715, CG17705, CG32143, CG32145, CG42280, CG9370, Dmel\CG42280, Dmel_CG42280, LD21715, Ome
Chromosome Location
Chr.3L: 14672839 - 14747868 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0259175
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144483.2 NP_001137955.1 2 - 2 929 67
GenBank mRNA BT024224.1 - 2 - 2 900 67

View Full Product Details