Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001103385.2 | NP_001096855.1 | 5 - 6 | 2139 | 78 |
NM_001103386.2 | NP_001096856.1 | 5 - 6 | 2139 | 78 | |
NM_001169155.1 | NP_001162626.1 | 4 - 5 | 2362 | 78 | |
NM_001297795.1 | NP_001284724.1 | 5 - 6 | 2140 | 78 | |
NM_001297796.1 | NP_001284725.1 | 5 - 6 | 2146 | 78 | |
NM_001297797.1 | NP_001284726.1 | 4 - 5 | 1932 | 78 | |
GenBank mRNA | BT024449.1 | - | 5 - 6 | 2101 | 78 |
BT081989.1 | - | 4 - 5 | 2362 | 78 |