Assay Design



Gene Transcripts

Gene Symbol
DIP-beta
Entrez Gene ID
33125
Gene Name
Dpr-interacting protein beta
Gene Aliases
42343, BP1062, BcDNA:RE23430, CG12564, CG12565, CG14583, CG34019, CG42343, CT34319, Dmel\CG42343, Dmel_CG42343, pp-CT34319, pp-CT34320, pp-CT34321
Chromosome Location
Chr.X: 21422115 - 21457738 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0259245
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144753.2 NP_001138225.1 9 - 10 1602 81
NM_001144754.2 NP_001138226.1 9 - 10 1517 81
NM_001298591.1 NP_001285520.1 9 - 10 1539 81

View Full Product Details