Assay Design
Gene Transcripts
- Gene Symbol
- CecA1
- Entrez Gene ID
-
43596
- Gene Name
- Cecropin A1
- Gene Aliases
-
CEC, CG1365, Cec, Cec A1, Cec-A1, CecA, DIM 25, Dmel\CG1365, Dmel_CG1365, ce-A1, cec, cecA, cecA1, crpA1
- Chromosome Location
-
Chr.3R: 30210874 - 30211273 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000276
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079849.4 |
NP_524588.1 |
2 - 2 |
251 |
60 |
View Full Product Details