Assay Design
Gene Transcripts
- Gene Symbol
- Gs1l
- Entrez Gene ID
-
33653
- Gene Name
- GS1-like
- Gene Aliases
-
19864674, CG15441, Dmel\CG15441, Dmel_CG15441, GS1l, gs1-like
- Chromosome Location
-
Chr.2L: 4455137 - 4456807 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0019982
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057881.5 |
NP_477229.3 |
2 - 3 |
213 |
65 |
View Full Product Details