Assay Design



Gene Transcripts

Gene Symbol
Haspin
Entrez Gene ID
3355064
Gene Name
CG40080-PA
Gene Aliases
BcDNA:LD07633, CG40080, Dmel\CG40080, Dmel_CG40080, haspin
Chromosome Location
Chr.2R: 4014111 - 4049342 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0046706
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015349.4 NP_001015349.2 2 - 3 1040 151
GenBank mRNA AY118897.1 - 2 - 3 1068 151

View Full Product Details