Assay Design



Gene Transcripts

Gene Symbol
Scp1
Entrez Gene ID
3355102
Gene Name
Sarcoplasmic calcium-binding protein 1
Gene Aliases
BcDNA:GH15296, CG15848, Dcabp-A.1, Dmel\CG15848, Dmel_CG15848, SCP
Chromosome Location
Chr.2R: 2932595 - 2968052 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020908
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015389.3 NP_001015389.1 4 - 5 369 149
NM_001276231.1 NP_001263160.1 4 - 5 410 149
GenBank mRNA AY118788.1 - 3 - 4 347 149

View Full Product Details