Assay Design
Gene Transcripts
- Gene Symbol
- Doa
- Entrez Gene ID
-
43415
- Gene Name
- Darkener of apricot
- Gene Aliases
-
0844/02, CG1658, CG31049, CG33204, CG33553, CG42320, DOA, DOA/CLK2, Dmel\CG42320, Dmel_CG42320, Msu, doa, l(3)01705, l(3)S084402, l(3)s2784
- Chromosome Location
-
Chr.3R: 28888073 - 28922772 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0265998
View Full Product Details