Assay Design



Gene Transcripts

Gene Symbol
adgrv1
Entrez Gene ID
415105
Gene Name
adhesion G protein-coupled receptor V1
Gene Aliases
gpr98, mass1, vlgr1
Chromosome Location
Chr.5: 48683336 - 48940174 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001845.2 NP_001001845.2 84 - 85 17980 87
XM_005165511.4 XP_005165568.1 84 - 85 18099 87
XM_009301753.3 XP_009300028.1 85 - 86 18121 87
GenBank mRNA AY540131.1 - 84 - 85 17984 87
BC090691.1 - 5 - 6 570 87

View Full Product Details