Assay Design



Gene Transcripts

Gene Symbol
cenpp
Entrez Gene ID
445161
Gene Name
centromere protein P
Gene Aliases
fb81h12, si:ch211-103f16.3, wu:fb81h12, zgc:101125
Chromosome Location
Chr.22: 10441004 - 10513965 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003555.3 NP_001003555.2 2 - 3 213 88
GenBank mRNA BC078335.1 - 2 - 3 187 88

View Full Product Details