Assay Design



Gene Transcripts

Gene Symbol
mapk7
Entrez Gene ID
541323
Gene Name
mitogen-activated protein kinase 7
Gene Aliases
bmk1, erk5, fb73b02, wu:fb73b02, zgc:113111
Chromosome Location
Chr.12: 4222840 - 4238827 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013451.3 NP_001013469.2 1 - 2 306 80
XM_005156002.4 XP_005156059.1 2 - 3 438 80
GenBank mRNA BC090470.1 - 2 - 3 575 80
DQ360072.1 - 1 - 2 306 80

View Full Product Details