Assay Design



Gene Transcripts

Gene Symbol
pdlim1
Entrez Gene ID
550568
Gene Name
PDZ and LIM domain 1 (elfin)
Gene Aliases
clim1, wu:fa19a03, wu:fy34f10, zgc:110681
Chromosome Location
Chr.13: 25762909 - 25773963 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017870.1 NP_001017870.1 3 - 4 399 108
GenBank mRNA BC092978.1 - 3 - 4 399 108

View Full Product Details