Assay Design



Gene Transcripts

Gene Symbol
crfb12
Entrez Gene ID
678605
Gene Name
cytokine receptor family member B12
Gene Aliases
zgc:136623
Chromosome Location
Chr.5: 57673140 - 57686485 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040353.1 NP_001035443.1 5 - 6 764 135
GenBank mRNA BC098871.1 - 5 - 6 766 135
BC115200.1 - 5 - 6 764 135

View Full Product Details