Assay Design



Gene Transcripts

Gene Symbol
daxx
Entrez Gene ID
561006
Gene Name
death-domain associated protein
Gene Aliases
si:dz179b20.7, wu:fd15c03
Chromosome Location
Chr.19: 7013053 - 7033667 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044949.1 NP_001038414.1 8 - 9 1464 178
XM_005159438.4 XP_005159495.2 8 - 9 1424 178
XM_005159440.4 XP_005159497.1 8 - 9 1448 178
XM_017352251.2 XP_017207740.1 8 - 9 1424 178

View Full Product Details