Assay Design



Gene Transcripts

Gene Symbol
plekhb2
Entrez Gene ID
100003431
Gene Name
pleckstrin homology domain containing, family B (evectins) member 2
Gene Aliases
si:dkey-121a11.4, zgc:55604
Chromosome Location
Chr.22: 16432934 - 16443153 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201002.1 NP_957296.1 3 - 4 266 73
GenBank mRNA BC045398.1 - 3 - 4 266 73
BC171510.1 - 3 - 4 242 73
BC171512.1 - 3 - 4 242 73

View Full Product Details