Assay Design



Gene Transcripts

Gene Symbol
bmi1b
Entrez Gene ID
394249
Gene Name
bmi1 polycomb ring finger oncogene 1b
Gene Aliases
pcgf4b
Chromosome Location
Chr.2: 21164930 - 21170269 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080751.1 NP_001074220.1 8 - 9 668 157
XM_021479461.1 XP_021335136.1 8 - 9 745 157
GenBank mRNA BC053151.1 - 8 - 9 668 157

View Full Product Details