Assay Design



Gene Transcripts

Gene Symbol
ube2l3b
Entrez Gene ID
321943
Gene Name
ubiquitin-conjugating enzyme E2L 3b
Gene Aliases
UBE2L3, ube2l3l, wu:fb38h03
Chromosome Location
Chr.5: 13337507 - 13353898 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082797.1 NP_001076266.1 1 - 2 320 122

View Full Product Details