Assay Design



Gene Transcripts

Gene Symbol
csnk2a1
Entrez Gene ID
30502
Gene Name
casein kinase 2, alpha 1 polypeptide
Gene Aliases
ck2a1, wu:fi38e04, wu:fi38h03
Chromosome Location
Chr.16: 303572 - 319194 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131252.1 NP_571327.1 6 - 7 594 97
XM_005158170.3 XP_005158227.1 6 - 7 642 97
GenBank mRNA BC044403.1 - 6 - 7 594 97
BC067595.1 - 5 - 6 511 97
BC165881.1 - 3 - 4 352 97
S76875.1 - 2 - 3 134 97

View Full Product Details