Assay Design



Gene Transcripts

Gene Symbol
msx1b
Entrez Gene ID
30511
Gene Name
muscle segment homeobox 1b
Gene Aliases
etID309829.1, msh-B, mshB, msxb, wu:fk04h02, zgc:110559
Chromosome Location
Chr.1: 49352935 - 49355307 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131260.1 NP_571335.1 1 - 2 423 84
GenBank mRNA BC095311.1 - 1 - 2 423 84
BC165505.1 - 1 - 2 403 84
U16311.1 - 1 - 2 359 84

View Full Product Details