Assay Design



Gene Transcripts

Gene Symbol
atp1b3b
Entrez Gene ID
64272
Gene Name
ATPase Na+/K+ transporting subunit beta 3b
Gene Aliases
cb714, fb13c07, wu:fb13c07
Chromosome Location
Chr.15: 4353163 - 4415803 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131670.1 NP_571745.1 1 - 2 184 121
GenBank mRNA AF293369.1 - 1 - 2 184 121
BC076055.1 - 1 - 2 148 121

View Full Product Details