Assay Design



Gene Transcripts

Gene Symbol
mib1
Entrez Gene ID
352910
Gene Name
mindbomb E3 ubiquitin protein ligase 1
Gene Aliases
cg5841, chunp6889, fe47f05, im:7148100, mib, wu:fe47f05
Chromosome Location
Chr.2: 4074171 - 4161272 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173286.3 NP_775393.2 4 - 5 731 97
XM_009296844.3 XP_009295119.1 4 - 5 1633 97
XM_009296845.3 XP_009295120.1 4 - 5 1633 97
XM_009296846.3 XP_009295121.1 4 - 5 1633 97
XM_009296848.3 XP_009295123.1 4 - 5 1633 97
XM_009296854.3 XP_009295129.1 4 - 5 1633 97
XM_009296859.3 XP_009295134.1 4 - 5 1633 97
GenBank mRNA AF506233.1 - 4 - 5 1392 97
AF537301.1 - 4 - 5 731 97

View Full Product Details