Assay Design



Gene Transcripts

Gene Symbol
mib1
Entrez Gene ID
352910
Gene Name
mindbomb E3 ubiquitin protein ligase 1
Gene Aliases
cg5841, chunp6889, fe47f05, im:7148100, mib, wu:fe47f05
Chromosome Location
Chr.2: 4074171 - 4161272 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173286.3 NP_775393.2 8 - 9 1336 71
XM_009296854.3 XP_009295129.1 8 - 9 2238 71
GenBank mRNA AF537301.1 - 8 - 9 1336 71

View Full Product Details