Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182877.2 | NP_878297.1 | 10 - 11 | 1277 | 86 |
XM_021469757.1 | XP_021325432.1 | 11 - 12 | 1727 | 86 | |
GenBank mRNA | AF297180.2 | - | 10 - 11 | 1278 | 86 |
BC054940.1 | - | 11 - 12 | 1296 | 86 |