Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199858.3 | NP_956152.1 | 5 - 6 | 687 | 66 |
XM_005156272.4 | XP_005156329.1 | 6 - 7 | 741 | 66 | |
XM_005156273.4 | XP_005156330.1 | 6 - 7 | 787 | 66 | |
GenBank mRNA | BC047189.1 | - | 6 - 7 | 722 | 66 |
BC065846.1 | - | 5 - 6 | 692 | 66 |