Assay Design



Gene Transcripts

Gene Symbol
fam49ba
Entrez Gene ID
337591
Gene Name
family with sequence similarity 49 member Ba
Gene Aliases
fam49b, fj85c11, wu:fj85c11, zgc:64118
Chromosome Location
Chr.2: 32215272 - 32262287 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291713.1 NP_001278642.1 5 - 6 833 77
NM_200070.3 NP_956364.2 5 - 6 833 77
GenBank mRNA AY394961.1 - 5 - 6 533 77
BC053261.1 - 5 - 6 519 77
EH507686.1 - 1 - 2 234 77

View Full Product Details