Assay Design



Gene Transcripts

Gene Symbol
fam49ba
Entrez Gene ID
337591
Gene Name
family with sequence similarity 49 member Ba
Gene Aliases
fam49b, fj85c11, wu:fj85c11, zgc:64118
Chromosome Location
Chr.2: 32215272 - 32262287 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200070.3 NP_956364.2 8 - 9 1109 65
GenBank mRNA AY394961.1 - 8 - 9 809 65
DT055468.1 - 3 - 4 673 65
EH507686.1 - 4 - 5 510 65

View Full Product Details