Assay Design



Gene Transcripts

Gene Symbol
jkamp
Entrez Gene ID
393454
Gene Name
jnk1/mapk8 associated membrane protein
Gene Aliases
zgc:63678, zgc:92212
Chromosome Location
Chr.17: 14443406 - 14451640 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200482.2 NP_956776.2 4 - 5 537 74
GenBank mRNA BC055200.1 - 4 - 5 353 74
BC075939.1 - 4 - 5 537 74

View Full Product Details