Assay Design
Gene Transcripts
- Gene Symbol
- fam3a
- Entrez Gene ID
-
393773
- Gene Name
- FAM3 metabolism regulating signaling molecule A
- Gene Aliases
-
zgc:73350
- Chromosome Location
-
Chr.23: 25135922 - 25150288 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200800.1 |
NP_957094.1 |
7 - 8 |
628 |
112 |
GenBank mRNA |
BC059665.1 |
- |
7 - 8 |
628 |
112 |
View Full Product Details