Assay Design
Gene Transcripts
- Gene Symbol
- tmem38a
- Entrez Gene ID
-
393874
- Gene Name
- transmembrane protein 38A
- Gene Aliases
-
zgc:77831
- Chromosome Location
-
Chr.11: 7158723 - 7179516 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200900.1 |
NP_957194.1 |
3 - 4 |
517 |
73 |
GenBank mRNA |
BC064309.1 |
- |
3 - 4 |
517 |
73 |
View Full Product Details