Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002650.1 | NP_001002650.1 | 4 - 5 | 882 | 65 |
XM_009298725.3 | XP_009297000.1 | 5 - 6 | 1216 | 65 | |
XM_017358225.2 | XP_017213714.1 | 5 - 6 | 1214 | 65 | |
GenBank mRNA | BC076526.1 | - | 4 - 5 | 912 | 65 |
BC164710.1 | - | 4 - 5 | 671 | 65 |