Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003889.1 | NP_001003889.1 | 5 - 6 | 576 | 71 |
GenBank mRNA | AY648806.1 | - | 5 - 6 | 576 | 71 |
BC085580.1 | - | 5 - 6 | 571 | 71 | |
BC165077.1 | - | 5 - 6 | 566 | 71 |