Assay Design



Gene Transcripts

Gene Symbol
ech1
Entrez Gene ID
450048
Gene Name
enoyl CoA hydratase 1, peroxisomal
Gene Aliases
zgc:101710
Chromosome Location
Chr.15: 13436 - 32066 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006068.1 NP_001006068.1 4 - 5 635 99
GenBank mRNA BC083254.1 - 4 - 5 635 99
BC152159.1 - 4 - 5 435 99
BC164085.1 - 4 - 5 453 99

View Full Product Details