Assay Design



Gene Transcripts

Gene Symbol
acot8
Entrez Gene ID
450052
Gene Name
acyl-CoA thioesterase 8
Gene Aliases
zgc:101691
Chromosome Location
Chr.11: 1512583 - 1520946 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006072.1 NP_001006072.1 2 - 3 280 74
GenBank mRNA BC083249.1 - 2 - 3 280 74
BC164121.1 - 2 - 3 305 74

View Full Product Details