Assay Design



Gene Transcripts

Gene Symbol
cyp1d1
Entrez Gene ID
492344
Gene Name
cytochrome P450, family 1, subfamily D, polypeptide 1
Gene Aliases
zgc:92205
Chromosome Location
Chr.5: 25606658 - 25621320 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007310.1 NP_001007311.1 2 - 3 951 73
GenBank mRNA BC085648.1 - 2 - 3 951 73
FJ416496.1 - 2 - 3 947 73

View Full Product Details