Assay Design



Gene Transcripts

Gene Symbol
extl3
Entrez Gene ID
493783
Gene Name
exostosin-like glycosyltransferase 3
Gene Aliases
im:7141548
Chromosome Location
Chr.20: 49940818 - 50014992 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008402.1 NP_001008402.1 2 - 3 2356 121
XM_005170211.4 XP_005170268.1 3 - 4 2866 121
GenBank mRNA AY786436.1 - 2 - 3 2356 121
BC162406.1 - 2 - 3 2347 121

View Full Product Details