Assay Design



Gene Transcripts

Gene Symbol
mmp23bb
Entrez Gene ID
550589
Gene Name
matrix metallopeptidase 23bb
Gene Aliases
mmp23al, mmp23b, zgc:110623
Chromosome Location
Chr.18: 45991028 - 46005525 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017890.1 NP_001017890.1 2 - 3 409 76
GenBank mRNA BC092954.1 - 2 - 3 409 76
BC164476.1 - 2 - 3 260 76

View Full Product Details