Assay Design



Gene Transcripts

Gene Symbol
mov10b.1
Entrez Gene ID
556024
Gene Name
Moloney leukemia virus 10b, tandem duplicate 1
Gene Aliases
fa99a10, mov10a, si:dkeyp-38g6.2, wu:fa99a10
Chromosome Location
Chr.8: 628065 - 655367 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044342.2 NP_001037807.2 6 - 7 1081 96
GenBank mRNA BC171375.1 - 6 - 7 1081 96

View Full Product Details