Assay Design



Gene Transcripts

Gene Symbol
cyp2k8
Entrez Gene ID
561462
Gene Name
cytochrome P450, family 2, subfamily K, polypeptide 8
Gene Aliases
zgc:194733
Chromosome Location
Chr.3: 12784460 - 12802380 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130623.2 NP_001124095.1 5 - 6 956 129
GenBank mRNA AY772712.1 - 5 - 6 872 129
BC163074.1 - 5 - 6 956 129

View Full Product Details