Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001089421.1 | NP_001082890.1 | 5 - 6 | 445 | 69 |
XM_005172951.4 | XP_005173008.1 | 4 - 5 | 796 | 69 | |
XM_021480155.1 | XP_021335830.1 | 5 - 6 | 833 | 69 | |
XM_021480156.1 | XP_021335831.1 | 4 - 5 | 801 | 69 | |
GenBank mRNA | AB438979.1 | - | 5 - 6 | 445 | 69 |
EF489398.1 | - | 5 - 6 | 445 | 69 |