Assay Design



Gene Transcripts

Gene Symbol
oxsr1a
Entrez Gene ID
568602
Gene Name
oxidative stress responsive kinase 1a
Gene Aliases
wu:fb77f12, zgc:165661
Chromosome Location
Chr.2: 23701659 - 23721336 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098747.1 NP_001092217.1 11 - 12 1222 73
GenBank mRNA BC124256.1 - 11 - 12 1269 73
BC142933.1 - 11 - 12 1222 73

View Full Product Details