Assay Design



Gene Transcripts

Gene Symbol
ankdd1a
Entrez Gene ID
569271
Gene Name
ankyrin repeat and death domain containing 1A
Gene Aliases
zgc:162892
Chromosome Location
Chr.7: 13582263 - 13618498 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099248.1 NP_001092718.1 12 - 13 1270 108
GenBank mRNA BC139890.1 - 12 - 13 1270 108

View Full Product Details