Assay Design



Gene Transcripts

Gene Symbol
ppm1aa
Entrez Gene ID
30704
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent, 1Aa
Gene Aliases
pp2ca2
Chromosome Location
Chr.13: 31545787 - 31580190 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131429.1 NP_571504.1 4 - 5 1418 97
XM_005156774.4 XP_005156831.1 4 - 5 1443 97
XM_005156775.4 XP_005156832.1 4 - 5 1444 97
XM_021480518.1 XP_021336193.1 4 - 5 1444 97
GenBank mRNA BC066510.1 - 4 - 5 1418 97

View Full Product Details