Assay Design
Gene Transcripts
- Gene Symbol
- ppm1aa
- Entrez Gene ID
-
30704
- Gene Name
- protein phosphatase, Mg2+/Mn2+ dependent, 1Aa
- Gene Aliases
-
pp2ca2
- Chromosome Location
-
Chr.13: 31545787 - 31580190 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_131429.1 |
NP_571504.1 |
5 - 6 |
1472 |
93 |
GenBank mRNA |
BC066510.1 |
- |
5 - 6 |
1472 |
93 |
View Full Product Details