Assay Design



Gene Transcripts

Gene Symbol
cyp19a1b
Entrez Gene ID
60640
Gene Name
cytochrome P450, family 19, subfamily A, polypeptide 1b
Gene Aliases
AroB, CYP19a2, P450aromB, cyp19, cyp19b, zgc:92614
Chromosome Location
Chr.25: 4890767 - 4904789 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131642.2 NP_571717.2 3 - 4 389 96
GenBank mRNA AF183908.1 - 3 - 4 368 96
AF226619.1 - 3 - 4 372 96
BC076104.1 - 3 - 4 386 96

View Full Product Details